Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EBLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018029.4 | NP_060499.3 | 1 | 734 | ||
GenBank mRNA | AK001075.1 | - | 1 | 121 | ||
AY036895.1 | - | 1 | 312 | |||
BC051316.1 | - | 1 | 734 | 6214587 | ||
BC064501.1 | - | 1 | 734 | 6215884 |