Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014245.4 | NP_055060.1 | 1, 2 | 309 | ||
GenBank mRNA | AF092878.1 | - | 1, 2 | 176 | ||
AF142060.1 | - | 1, 2 | 176 | |||
AF164679.1 | - | 1, 2 | 229 | |||
AK311982.1 | - | 1, 2 | 241 | |||
BC005966.1 | - | 1, 2 | 202 | 4069078 | ||
BC008627.1 | - | 1, 2 | 212 | 4177613 | ||
BG742338.1 | - | 1, 2 | 309 | |||
BT007348.1 | - | 1, 2 | 176 |