Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5SL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167867.1 | NP_001161339.1 | 2 | 291 | ||
NM_001167868.1 | NP_001161340.1 | 2 | 291 | |||
NM_001167869.1 | NP_001161341.1 | 2 | 245 | |||
NM_001320838.1 | NP_001307767.1 | 2 | 245 | |||
NM_018035.2 | NP_060505.2 | 2 | 245 | |||
NR_135476.1 | 2 | 245 | ||||
NR_135478.1 | 2 | 245 | ||||
XM_011527065.2 | XP_011525367.1 | 2 | 296 | |||
GenBank mRNA | AK001103.1 | - | 2 | 212 | ||
AK293132.1 | - | 2 | 254 | |||
AK297698.1 | - | 2 | 215 | |||
AK301940.1 | - | 2 | 254 | |||
DA057313.1 | - | 2 | 245 | |||
DA075924.1 | - | 2 | 245 | |||
DA540661.1 | - | 2 | 213 | |||
DA631852.1 | - | 2 | 215 |