Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017999.2 | NP_001017999.1 | 7 | 1114 | ||
NM_001018000.3 | NP_001018000.1 | 7 | 1303 | |||
NM_001018001.2 | NP_001018001.1 | 7 | 1104 | |||
NM_015209.2 | NP_056024.1 | 7 | 1366 | |||
NM_201628.2 | NP_963922.2 | 7 | 1366 | |||
XM_005245795.4 | XP_005245852.1 | 9 | 1621 | |||
XM_011541074.2 | XP_011539376.1 | 9 | 1628 | |||
XM_011541075.2 | XP_011539377.1 | 7 | 1284 | |||
XM_011541076.2 | XP_011539378.1 | 7 | 1429 | |||
XM_011541077.2 | XP_011539379.1 | 7 | 2408 | |||
XM_011541080.2 | XP_011539382.1 | 9 | 1621 | |||
XM_017000768.1 | XP_016856257.1 | 9 | 1621 | |||
XM_017000769.1 | XP_016856258.1 | 9 | 1616 | |||
XM_017000770.1 | XP_016856259.1 | 9 | 1616 | |||
XM_017000771.1 | XP_016856260.1 | 8 | 1289 | |||
XM_017000772.1 | XP_016856261.1 | 7 | 968 | |||
GenBank mRNA | AB015329.1 | - | 6 | 756 | ||
AK293806.1 | - | 2 | 172 | |||
AY505119.1 | - | 8 | 1294 | |||
AY505120.1 | - | 7 | 1303 | |||
AY505121.1 | - | 7 | 1104 | |||
AY505122.1 | - | 7 | 1114 | |||
BC035501.2 | - | 7 | 1302 | 4556194 | ||
DR003677.1 | - | 2 | 89 |