Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 4513 | ||
NM_001160258.1 | NP_001153730.1 | 4 | 4551 | |||
NM_001160259.1 | NP_001153731.1 | 2 | 4426 | |||
NM_016083.4 | NP_057167.2 | 2 | 4370 | |||
NM_033181.3 | NP_149421.2 | 2 | 4025 | |||
XM_006715330.3 | XP_006715393.1 | 2 | 4317 | |||
XM_011535424.1 | XP_011533726.1 | 3 | 4857 | |||
XM_011535425.1 | XP_011533727.1 | 4 | 4895 | |||
XM_011535426.1 | XP_011533728.1 | 5 | 5053 | |||
XM_011535428.1 | XP_011533730.1 | 3 | 4704 | |||
XM_017010238.1 | XP_016865727.1 | 4 | 7681 | |||
XM_017010239.1 | XP_016865728.1 | 3 | 6004 | |||
XM_017010240.1 | XP_016865729.1 | 2 | 4364 | |||
GenBank mRNA | BX538191.1 | - | 1 | 1734 |
Set Membership: |
G-protein coupled receptor |