Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016806.3 | NP_058086.2 | 4 | 562 | ||
NM_182650.4 | NP_872591.1 | 4 | 562 | |||
GenBank mRNA | AF073993.1 | - | 4 | 440 | ||
AF406651.2 | - | 3 | 351 | |||
AK049771.1 | - | 3 | 2927 | |||
AK089012.1 | - | 4 | 513 | |||
AK137214.1 | - | 4 | 563 | |||
BB627924.1 | - | 4 | 563 | |||
BC059107.1 | - | 2 | 98 | |||
BC141253.1 | - | 4 | 396 | 9055865 | ||
BC145588.1 | - | 4 | 396 | 9054154 |