Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026241.3 | NP_080517.1 | 4 | 1361 | ||
GenBank mRNA | AK018681.1 | - | 4 | 1362 | ||
AK030874.1 | - | 1557 | ||||
AK042896.1 | - | 4 | 1362 | |||
AK158215.1 | - | 4 | 1557 |