Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSEN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000021.3 | NP_000012.1 | 4, 5 | 619 | ||
NM_007318.2 | NP_015557.2 | 4, 5 | 607 | |||
XM_005267864.2 | XP_005267921.1 | 4, 5 | 613 | |||
XM_005267866.1 | XP_005267923.1 | 4, 5 | 602 | |||
XM_011536971.2 | XP_011535273.1 | 4, 5 | 511 | |||
XM_011536972.2 | XP_011535274.1 | 4, 5 | 875 | |||
XM_011536973.1 | XP_011535275.1 | 4, 5 | 863 | |||
XM_011536974.1 | XP_011535276.1 | 4, 5 | 504 | |||
GenBank mRNA | AF416717.1 | - | 2, 3 | 335 | ||
AJ008005.1 | - | 4, 5 | 553 | |||
AK122722.1 | - | 2, 3 | 1888 | |||
AK223173.1 | - | 2, 3 | 349 | |||
AK312531.1 | - | 4, 5 | 603 | |||
BC011729.2 | - | 4, 5 | 588 | 4423098 | ||
KU178285.1 | - | 2, 3 | 323 | |||
KU178286.1 | - | 2, 3 | 335 | |||
L42110.1 | - | 4, 5 | 583 | |||
L76517.1 | - | 4, 5 | 876 | |||
U40379.1 | - | 2, 3 | 323 | |||
U40380.1 | - | 2, 3 | 323 |