Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027560.1 | NP_081836.1 | 7 | 1414 | ||
GenBank mRNA | AK014582.1 | - | 7 | 1414 | ||
AK046754.1 | - | 7 | 1124 | |||
AK135740.1 | - | 7 | 1230 | |||
AK137175.1 | - | 7 | 1231 | |||
AK144323.1 | - | 8 | 1148 | |||
BC034910.1 | - | 1 | 293 | |||
BC036384.1 | - | 7 | 1212 | 1067080 | ||
BC043313.1 | - | 2 | 934 | |||
BC043469.1 | - | 2 | 934 | |||
BC054826.1 | - | 2 | 940 | |||
BC094060.1 | - | 6 | 1294 | |||
DQ861995.1 | - | 7 | 1078 |