Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027959.3 | NP_082235.1 | 13 | 1540 | ||
GenBank mRNA | AK076558.1 | - | 13 | 1541 | ||
AK133792.1 | - | 13 | 1548 | |||
AK148928.1 | - | 1989 | ||||
AK151008.1 | - | 13 | 1520 | |||
AK152960.1 | - | 13 | 1553 | |||
AK165877.1 | - | 13 | 1519 | |||
AK166111.1 | - | 13 | 1547 | |||
AK167386.1 | - | 13 | 1558 | |||
AK168136.1 | - | 13 | 1547 | |||
AK168279.1 | - | 13 | 1547 | |||
BC006865.1 | - | 13 | 1529 | 2645183 |
Gene Symbol: LOC102642954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK206634.1 | - | 1 | 73 |