Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030017.3 | NP_084293.1 | 7 | 1165 | ||
GenBank mRNA | AK039233.1 | - | 7 | 1166 | ||
BC016204.1 | - | 7 | 1112 | 4527851 | ||
BC067213.1 | - | 6 | 2286 |