Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM86B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137610.1 | NP_001131082.1 | 8 | 1154 | ||
XM_005273603.4 | XP_005273660.1 | 7 | 1127 | |||
XM_005273604.4 | XP_005273661.1 | 6 | 1046 | |||
XM_005273605.4 | XP_005273662.1 | 6 | 947 | |||
XM_005273607.2 | XP_005273664.1 | 6 | 985 | |||
XM_006716387.3 | XP_006716450.1 | 7 | 1148 | |||
XM_011544618.2 | XP_011542920.1 | 8 | 1199 | |||
XM_011544619.2 | XP_011542921.1 | 7 | 1166 | |||
XM_011544622.2 | XP_011542924.1 | 7 | 1068 | |||
XM_017013740.1 | XP_016869229.1 | 6 | 1129 | |||
XM_017013741.1 | XP_016869230.1 | 5 | 1066 | |||
XM_017013742.1 | XP_016869231.1 | 5 | 1048 | |||
GenBank mRNA | AK297741.1 | - | 5 | 729 | ||
AK298940.1 | - | 4 | 847 |
Gene Symbol: FAM86B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083537.1 | NP_001077006.1 | 7 | 1097 | ||
NR_003494.1 | 6 | 831 | ||||
XM_006716256.3 | XP_006716319.1 | 8 | 1192 | |||
XM_006716257.3 | XP_006716320.1 | 7 | 1262 | |||
XM_006716260.3 | XP_006716323.1 | 6 | 1006 | |||
XM_006716262.1 | XP_006716325.1 | 6 | 849 | |||
XM_011543842.2 | XP_011542144.1 | 6 | 1271 | |||
XM_011543843.2 | XP_011542145.1 | 7 | 1210 | |||
XM_017013916.1 | XP_016869405.1 | 7 | 1238 | |||
XM_017013917.1 | XP_016869406.1 | 6 | 1159 | |||
XM_017013918.1 | XP_016869407.1 | 6 | 1131 | |||
XM_017013919.1 | XP_016869408.1 | 5 | 720 | |||
GenBank mRNA | AK098138.1 | - | 6 | 798 | ||
AK308439.1 | - | 7 | 912 | |||
AL529666.3 | - | 5 | 768 | |||
BC062615.1 | - | 6 | 875 | |||
BC085016.1 | - | 7 | 904 | |||
BC098427.1 | - | 6 | 881 |