Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phyhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252568.2 | NP_001239497.1 | 11 | 1488 | ||
NM_001252570.1 | NP_001239499.1 | 11 | 1178 | |||
NM_001252571.1 | NP_001239500.1 | 10 | 1115 | |||
NM_001281829.1 | NP_001268758.1 | 12 | 1204 | |||
NM_172267.4 | NP_758471.1 | 12 | 1557 | |||
GenBank mRNA | AK005293.1 | - | 12 | 1558 | ||
AK138607.1 | - | 12 | 1232 | |||
AK193937.1 | - | 1 | 138 | |||
BC039982.1 | - | 12 | 1199 | |||
BU701903.1 | - | 2 | 340 |