Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175535.3 | NP_780744.2 | 15 | 2849 | ||
GenBank mRNA | AK129345.1 | - | 12 | 2338 |