Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175843.4 | NP_787037.2 | 3, 4 | 555 | ||
NM_181550.1 | NP_853528.1 | 3, 4 | 521 | |||
XM_006246213.2 | XP_006246275.1 | 3, 4 | 577 | |||
XM_006246214.2 | XP_006246276.1 | 3, 4 | 577 | |||
XM_006246215.2 | XP_006246277.1 | 3, 4 | 577 | |||
GenBank mRNA | AF439403.1 | - | 3, 4 | 521 | ||
BC061575.1 | - | 3, 4 | 559 | 6887571 | ||
Y08355.1 | - | 3, 4 | 584 |