Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177905.3 | NP_808573.2 | 14 | 1948 | ||
GenBank mRNA | AB258534.1 | - | 14 | 1906 | ||
AK143022.1 | - | 5 | 718 |