Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318716.1 | NP_001305645.1 | 2 | 616 | ||
NM_001318717.1 | NP_001305646.1 | 2 | 537 | |||
NM_001318718.1 | NP_001305647.1 | 2 | 517 | |||
NM_001318719.1 | NP_001305648.1 | 2 | 495 | |||
NM_001318720.1 | NP_001305649.1 | 2 | 489 | |||
NM_001318721.1 | NP_001305650.1 | 2 | 464 | |||
NM_001318722.1 | NP_001305651.1 | 2 | 485 | |||
NM_001318723.1 | NP_001305652.1 | 2 | 569 | |||
NM_001318724.1 | NP_001305653.1 | 2 | 569 | |||
NM_001318725.1 | NP_001305654.1 | 2 | 472 | |||
NM_016506.6 | NP_057590.3 | 2 | 573 | |||
NM_018095.5 | NP_060565.4 | 2 | 468 | |||
XM_017018009.1 | XP_016873498.1 | 2 | 452 | |||
GenBank mRNA | AK001312.1 | - | 2 | 422 | ||
AK001749.1 | - | 2 | 509 | |||
AK091607.1 | - | 1 | 1146 | |||
AK222550.1 | - | 2 | 419 | |||
AK222985.1 | - | 2 | 429 | |||
AK225062.1 | - | 2 | 419 | |||
AL713735.1 | - | 1 | 398 | |||
BC002736.2 | - | 2 | 510 | 3628290 | ||
BI601113.1 | - | 2 | 476 | |||
BI908078.1 | - | 2 | 475 | |||
CR457270.1 | - | 1 | 356 | |||
DA016320.1 | - | 2 | 468 | |||
DA883418.1 | - | 2 | 506 | |||
DB457928.1 | - | 2 | 440 | |||
HY062454.1 | - | 2 | 451 | |||
HY120885.1 | - | 2 | 450 | |||
HY176105.1 | - | 2 | 492 |