Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fam89b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023166.2 | NP_075655.2 | 2 | 692 | ||
NM_181452.2 | NP_852117.2 | 2 | 731 | |||
GenBank mRNA | AB012726.1 | - | 2 | 716 | ||
AF212920.1 | - | 2 | 713 | |||
AF212921.1 | - | 2 | 659 | |||
AK003883.1 | - | 2 | 681 | |||
AK013256.1 | - | 2 | 660 | |||
BC013894.1 | - | 2 | 719 | 4217979 | ||
BC056166.1 | - | 2 | 677 | 5715229 |