Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134479.1 | NP_001127951.1 | 3 | 1270 | ||
NM_018103.4 | NP_060573.2 | 3 | 1257 | |||
XM_011541689.1 | XP_011539991.1 | 4 | 1341 | |||
XM_017001599.1 | XP_016857088.1 | 3 | 1612 | |||
XM_017001600.1 | XP_016857089.1 | 3 | 1067 | |||
XM_017001601.1 | XP_016857090.1 | 3 | 1958 | |||
XM_017001602.1 | XP_016857091.1 | 3 | 1311 | |||
GenBank mRNA | AK001332.1 | - | 1 | 323 | ||
AK092238.1 | - | 3 | 1233 | |||
AK222787.1 | - | 1 | 284 | |||
AL831880.1 | - | 1 | 729 | |||
AL832952.1 | - | 1 | 688 | |||
BC009486.1 | - | 3 | 1202 | |||
BC024159.2 | - | 3 | 1224 | 3958069 |