Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xkrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012230.1 | NP_001012230.1 | 3 | 1618 | ||
GenBank mRNA | AY534257.1 | - | 3 | 1618 |