Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018004.2 | NP_060474.1 | 6 | 1247 | ||
XM_005247569.2 | XP_005247626.1 | 7 | 1208 | |||
XM_017006715.1 | XP_016862204.1 | 7 | 1368 | |||
GenBank mRNA | AK000996.1 | - | 6 | 1247 | ||
AK129789.1 | - | 6 | 1097 | |||
AY450392.1 | - | 6 | 1247 | |||
BC040355.1 | - | 7 | 1181 | 4748104 | ||
BG205309.1 | - | 2 | 349 | |||
BX648832.1 | - | 3 | 4301 |