Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025134.1 | NP_001020305.1 | 2 | 986 | ||
GenBank mRNA | AY323237.1 | - | 2 | 986 |