Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUC7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320226.1 | NP_001307155.1 | 4, 5 | 517 | ||
NM_018032.4 | NP_060502.1 | 4, 5 | 517 | |||
NM_201412.2 | NP_958815.1 | 4, 5 | 517 | |||
XM_005255427.3 | XP_005255484.1 | 5, 6 | 554 | |||
XM_005255429.3 | XP_005255486.1 | 6, 7 | 600 | |||
XM_011522561.2 | XP_011520863.1 | 3, 4 | 356 | |||
XM_017023437.1 | XP_016878926.1 | 4, 5 | 1580 | |||
XM_017023438.1 | XP_016878927.1 | 5, 6 | 1644 | |||
XM_017023439.1 | XP_016878928.1 | 4, 5 | 1580 | |||
XM_017023440.1 | XP_016878929.1 | 5, 6 | 1644 | |||
GenBank mRNA | AJ404326.1 | - | 4, 5 | 445 | ||
AK001093.1 | - | 4, 5 | 445 | |||
AK097974.1 | - | 2, 3 | 1295 | |||
AK223084.1 | - | 4, 5 | 448 | |||
AY005111.1 | - | 4, 5 | 485 | |||
BC003194.1 | - | 4, 5 | 462 | 3536109 | ||
BC065198.1 | - | 4, 5 | 1734 | 5212471 | ||
BX647161.1 | - | 4, 5 | 1515 |