Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028481.1 | NP_082757.1 | 11, 12 | 1582 | ||
GenBank mRNA | AK016519.1 | - | 11, 12 | 1576 | ||
AK040158.1 | - | 10, 11 | 1498 | |||
BC082598.1 | - | 11, 12 | 1582 | 30641632 |