Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028481.1 | NP_082757.1 | 18 | 2499 | ||
GenBank mRNA | AK016519.1 | - | 18 | 2493 | ||
BC082598.1 | - | 18 | 2499 | 30641632 |