Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028758.2 | NP_083034.1 | 9 | 975 | ||
GenBank mRNA | AK004632.1 | - | 9 | 947 | ||
AK034612.1 | - | 6 | 618 | |||
AK129280.1 | - | 9 | 934 | |||
AK143691.1 | - | 9 | 963 | |||
AK149798.1 | - | 9 | 965 | |||
AK154904.1 | - | 9 | 950 | |||
AK159953.1 | - | 9 | 965 | |||
BC057377.1 | - | 2 | 478 | 5698559 | ||
BC062933.1 | - | 9 | 944 | 30356060 |