Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028862.3 | NP_083138.2 | 11 | 2581 | ||
GenBank mRNA | AK014408.1 | - | 11 | 2582 | ||
AK043002.1 | - | 11 | 2568 | |||
AK196353.1 | - | 1 | 100 | |||
BC051064.1 | - | 4 | 1284 | |||
BC055485.1 | - | 5 | 1365 |