Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166553.1 | NP_001160025.1 | 2 | 454 | ||
NM_028862.3 | NP_083138.2 | 2 | 473 | |||
GenBank mRNA | AK014408.1 | - | 2 | 474 | ||
AK043002.1 | - | 2 | 460 | |||
AK087359.1 | - | 2 | 390 | |||
AK139310.1 | - | 2 | 371 | |||
BB618481.1 | - | 2 | 340 | |||
BC040799.1 | - | 1 | 149 | 3988157 |