Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028881.2 | NP_083157.1 | 14 | 2326 | ||
GenBank mRNA | AK014553.1 | - | 8 | 2433 | ||
AK079382.1 | - | 11 | 2667 | |||
AK156192.1 | - | 14 | 2305 | |||
AK169944.1 | - | 10 | 1975 | |||
BC019713.1 | - | 4 | 708 | |||
BC023091.1 | - | 10 | 1916 | 4023064 | ||
BC023831.1 | - | 14 | 2321 | 5345301 | ||
BC032183.1 | - | 14 | 2331 | 5310152 | ||
BC033300.1 | - | 14 | 2321 | 4166411 |