Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam228a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029107.2 | NP_083383.1 | 8 | 1714 | ||
GenBank mRNA | AK029492.1 | - | 8 | 1715 | ||
AK133272.1 | - | 8 | 1752 | |||
BC138846.1 | - | 8 | 1649 | 8861868 | ||
BC138853.1 | - | 8 | 1557 | 8861875 |