Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018052.3 | NP_060522.3 | 19 | 2537 | ||
XM_005256038.3 | XP_005256095.1 | 19 | 6507 | |||
GenBank mRNA | AK001167.1 | - | 9 | 996 | ||
AK025479.1 | - | 13 | 1570 | |||
AK056433.1 | - | 19 | 2547 | |||
AK090627.1 | - | 1 | 1730 | |||
AK093941.1 | - | 6 | 1516 | |||
AK128439.1 | - | 1 | 3052 | |||
AK297564.1 | - | 19 | 2322 | |||
AK307766.1 | - | 18 | 2387 | |||
AL137396.1 | - | 3 | 328 | |||
BC000536.2 | - | 9 | 1001 | |||
BC007214.1 | - | 14 | 1659 | |||
BC110852.1 | - | 14 | 1592 | |||
BC125108.1 | - | 19 | 2469 | 40118772 | ||
BC125109.1 | - | 19 | 2469 | 40118777 | ||
CR457343.1 | - | 13 | 1505 |