Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008506.2 | NP_001008506.2 | 6 | 966 | ||
NM_001160107.1 | NP_001153579.1 | 6 | 966 | |||
NM_029334.2 | NP_083610.2 | 6 | 966 | |||
GenBank mRNA | AK048046.1 | - | 6 | 932 | ||
AK050226.1 | - | 6 | 947 | |||
AK076066.1 | - | 5 | 847 | |||
AK160937.1 | - | 6 | 952 | |||
AK177154.1 | - | 1 | 106 | |||
AK193300.1 | - | 1 | 105 | |||
AK196806.1 | - | 1 | 105 | |||
AK197684.1 | - | 1 | 105 | |||
AK202592.1 | - | 1 | 105 | |||
AK202852.1 | - | 1 | 106 | |||
AK204578.1 | - | 1 | 105 | |||
AK207425.1 | - | 1 | 105 | |||
BC021797.1 | - | 2 | 398 | |||
BC024824.1 | - | 6 | 936 | 5355748 | ||
BC026610.1 | - | 2 | 431 | |||
BC034760.1 | - | 6 | 899 |