Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pifo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200028.1 | NP_001186957.1 | 1 | 191 | ||
GenBank mRNA | BC049757.1 | - | 1 | 192 | 6773491 | |
HM237138.1 | - | 1 | 191 |