Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278529.1 | NP_001265458.1 | 24 | 5150 | ||
NM_001278530.1 | NP_001265459.1 | 23 | 4812 | |||
NM_018071.4 | NP_060541.3 | 24 | 5123 | |||
XM_005267844.2 | XP_005267901.1 | 23 | 5015 | |||
XM_011536937.2 | XP_011535239.1 | 24 | 5171 | |||
XM_017021434.1 | XP_016876923.1 | 25 | 5428 | |||
XM_017021435.1 | XP_016876924.1 | 23 | 5645 | |||
XM_017021436.1 | XP_016876925.1 | 20 | 3428 | |||
XM_017021437.1 | XP_016876926.1 | 19 | 3272 | |||
GenBank mRNA | AK001219.1 | - | 8 | 1329 | ||
AK024463.1 | - | 22 | 4361 | |||
AK074057.1 | - | 24 | 5068 | |||
AL137291.1 | - | 10 | 2817 | |||
BC000084.2 | - | 10 | 1673 | |||
BC142692.1 | - | 24 | 5093 | 40148708 | ||
BC151212.1 | - | 24 | 5093 | |||
BF339938.1 | - | 1 | 356 | |||
T97121.1 | - | 1 | 85 |