Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NECAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145277.1 | NP_001138749.1 | 7 | 1799 | ||
NM_001145278.1 | NP_001138750.1 | 8 | 1797 | |||
NM_018090.4 | NP_060560.1 | 8 | 1875 | |||
XM_011541767.2 | XP_011540069.1 | 8 | 1871 | |||
XM_017001721.1 | XP_016857210.1 | 7 | 1668 | |||
GenBank mRNA | AK001282.1 | - | 9 | 1596 | ||
AK021938.1 | - | 4 | 1351 | |||
AK023545.1 | - | 7 | 1735 | |||
AK024468.1 | - | 7 | 4427 | |||
AK223344.1 | - | 9 | 1097 | |||
BC017014.1 | - | 8 | 1806 | 3848563 | ||
BC018914.2 | - | 8 | 1809 | 4299890 |