Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134479.1 | NP_001127951.1 | 3 | 2965 | ||
NM_018103.4 | NP_060573.2 | 3 | 2952 | |||
XM_011541689.1 | XP_011539991.1 | 4 | 3036 | |||
XM_017001599.1 | XP_016857088.1 | 3 | 3307 | |||
XM_017001600.1 | XP_016857089.1 | 3 | 2762 | |||
XM_017001601.1 | XP_016857090.1 | 3 | 3653 | |||
XM_017001602.1 | XP_016857091.1 | 3 | 3006 | |||
GenBank mRNA | AK001332.1 | - | 1 | 2018 | ||
AK092238.1 | - | 3 | 2928 | |||
AK222787.1 | - | 1 | 1979 | |||
AL831880.1 | - | 1 | 2424 | |||
AL832952.1 | - | 1 | 2383 | |||
AY007147.1 | - | 1 | 721 | |||
AY358874.1 | - | 1 | 1540 | |||
BC024159.2 | - | 3 | 2919 | 3958069 |