Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF011415.1 | - | 2826 |
Gene Symbol: Vmn2r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009493.2 | NP_033519.1 | 6 | 3330 | ||
GenBank mRNA | AF011414.1 | - | 6 | 3332 |
Set Membership: |
G-protein coupled receptor |