Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cplx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009946.3 | NP_034076.1 | 2 | 322 | ||
GenBank mRNA | BC048487.1 | - | 2 | 192 | ||
CX241609.1 | - | 2 | 322 | |||
D38613.1 | - | 1 | 91 |