Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015378.3 | NP_056193.2 | 30 | 7286 | ||
NM_018156.3 | NP_060626.2 | 30 | 7286 | |||
GenBank mRNA | AB007922.3 | - | 12 | 3596 | ||
AJ608774.1 | - | 30 | 7258 | |||
AJ608775.1 | - | 30 | 7258 | |||
AK001481.1 | - | 3 | 406 | |||
AK125118.1 | - | 6 | 1016 | |||
BX641035.1 | - | 5 | 850 |