Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153151.3 | NP_694791.2 | 1, 2 | 151 | ||
GenBank mRNA | AF356876.1 | - | 1, 2 | 157 | ||
AK143884.1 | - | 1, 2 | 111 | |||
AK149441.1 | - | 1, 2 | 151 | |||
BC049873.1 | - | 1, 2 | 56 | 5054536 |