Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007596.2 | NP_001007597.1 | 7 | 1367 | ||
NM_001286448.1 | NP_001273377.1 | 7 | 1912 | |||
NM_153457.7 | NP_703187.2 | 9 | 3448 | |||
GenBank mRNA | BC027091.1 | - | 7 | 1247 | ||
BY615773.1 | - | 1 | 211 |