Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018197.2 | NP_060667.2 | 3 | 785 | ||
NM_199426.1 | NP_955458.1 | 3 | 779 | |||
NM_199427.2 | NP_955459.2 | 3 | 785 | |||
XM_005260449.1 | XP_005260506.1 | 2 | 202 | |||
XM_011528900.1 | XP_011527202.1 | 4 | 676 | |||
XM_017027945.1 | XP_016883434.1 | 3 | 571 | |||
XM_017027948.1 | XP_016883437.1 | 3 | 324 | |||
XM_017027949.1 | XP_016883438.1 | 4 | 643 | |||
GenBank mRNA | AK001596.1 | - | 3 | 575 | ||
AK090547.1 | - | 4 | 447 | |||
AK222712.1 | - | 3 | 579 | |||
BC012759.2 | - | 3 | 785 | 3634346 | ||
BC021087.1 | - | 3 | 506 | |||
BC041622.1 | - | 3 | 785 | 3633938 | ||
BT009760.1 | - | 3 | 436 |