Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243342.1 | NP_001230271.1 | 14 | 2466 | ||
NM_017950.3 | NP_060420.2 | 14 | 2466 | |||
XM_011524963.2 | XP_011523265.1 | 14 | 2503 | |||
XM_011524964.2 | XP_011523266.1 | 8 | 1435 | |||
GenBank mRNA | AB046860.1 | - | 10 | 1972 | ||
AK056583.1 | - | 3 | 845 | |||
AL713758.1 | - | 3 | 1062 | |||
BC035251.1 | - | 14 | 2459 | 4838111 |