Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018206.5 | NP_060676.2 | 2 | 179 | ||
XM_005256045.3 | XP_005256102.1 | 2 | 141 | |||
XM_011523227.2 | XP_011521529.1 | 2 | 909 | |||
GenBank mRNA | AF175265.1 | - | 2 | 137 | ||
AF183418.1 | - | 1 | 88 | |||
AF186382.1 | - | 2 | 133 | |||
AF191298.2 | - | 2 | 107 | |||
AK001614.1 | - | 2 | 127 | |||
AK223218.1 | - | 2 | 98 | |||
AL136888.1 | - | 2 | 127 | |||
BC002414.2 | - | 1 | 86 | 3162255 | ||
BC010362.1 | - | 2 | 100 | 4249949 | ||
BC093036.1 | - | 2 | 125 | 30340379 | ||
DB049686.1 | - | 2 | 179 |