Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173615.4 | NP_775886.3 | 34 | 3695 | ||
XM_011545742.2 | XP_011544044.1 | 38 | 3977 | |||
XM_011545744.2 | XP_011544046.1 | 23 | 2577 | |||
XM_011545745.1 | XP_011544047.1 | 21 | 2345 | |||
XM_011545746.2 | XP_011544048.1 | 21 | 2448 | |||
XM_017022947.1 | XP_016878436.1 | 39 | 4050 | |||
XM_017022948.1 | XP_016878437.1 | 22 | 3149 | |||
XM_017022949.1 | XP_016878438.1 | 20 | 3420 | |||
GenBank mRNA | AK093467.1 | - | 10 | 1258 | ||
AK128606.1 | - | 23 | 2563 | |||
BC038400.1 | - | 11 | 1140 | |||
BC109294.1 | - | 23 | 2470 | |||
BC109295.1 | - | 22 | 2423 |