Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170585.1 | NP_001164056.1 | 4 | 275 | ||
NM_153021.4 | NP_694566.4 | 4 | 275 | |||
XM_011532579.2 | XP_011530881.1 | 5 | 522 | |||
XM_011532581.2 | XP_011530883.1 | 5 | 518 | |||
XM_011532584.2 | XP_011530886.1 | 5 | 522 | |||
XM_011532588.2 | XP_011530890.1 | 5 | 524 | |||
XM_011532589.2 | XP_011530891.1 | 5 | 519 | |||
XM_011532590.2 | XP_011530892.1 | 5 | 524 | |||
XM_011532591.2 | XP_011530893.1 | 5 | 521 | |||
XM_011532606.2 | XP_011530908.1 | 5 | 544 | |||
XM_011532607.2 | XP_011530909.1 | 5 | 544 | |||
XM_011532608.2 | XP_011530910.1 | 5 | 542 | |||
XM_017003432.1 | XP_016858921.1 | 5 | 518 | |||
XM_017003433.1 | XP_016858922.1 | 5 | 517 | |||
XM_017003434.1 | XP_016858923.1 | 5 | 526 | |||
GenBank mRNA | BC150633.1 | - | 4 | 275 | 9057003 | |
BC153864.1 | - | 4 | 227 |