Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080504.2 | NP_001073973.2 | 6, 7 | 2239 | ||
XM_005246075.1 | XP_005246132.1 | 6, 7 | 2580 | |||
XM_011511152.2 | XP_011509454.1 | 6, 7 | 2580 | |||
XM_011511153.2 | XP_011509455.1 | 6, 7 | 2580 | |||
XM_011511154.2 | XP_011509456.1 | 6, 7 | 2580 | |||
XM_017004054.1 | XP_016859543.1 | 6, 7 | 2580 | |||
XM_017004055.1 | XP_016859544.1 | 6, 7 | 2950 | |||
XM_017004056.1 | XP_016859545.1 | 6, 7 | 2520 | |||
XM_017004057.1 | XP_016859546.1 | 6, 7 | 2580 | |||
XM_017004058.1 | XP_016859547.1 | 1, 2 | 918 | |||
GenBank mRNA | BC126111.1 | - | 3, 4 | 322 | ||
BX648107.1 | - | 5, 6 | 2892 |