Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000811.2 | NP_000802.2 | 2, 3 | 503 | ||
GenBank mRNA | AK289461.1 | - | 2, 3 | 233 | ||
BC096241.3 | - | 2, 3 | 191 | 40005676 | ||
BC096242.3 | - | 2, 3 | 191 | 40005680 | ||
BC099640.3 | - | 2, 3 | 191 | 40005678 | ||
BC099641.3 | - | 2, 3 | 191 | 40005679 | ||
S81944.1 | - | 2, 3 | 191 |