Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039111.2 | NP_001034200.1 | 4 | 1700 | ||
GenBank mRNA | DQ232881.1 | - | 4 | 1700 |