Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100159.2 | NP_001093629.1 | 5 | 706 | ||
NM_001267865.1 | NP_001254794.1 | 4 | 550 | |||
NM_001267866.1 | NP_001254795.1 | 4 | 550 | |||
XM_006715649.3 | XP_006715712.1 | 4 | 566 | |||
XM_006715650.2 | XP_006715713.1 | 4 | 533 | |||
XM_011515121.1 | XP_011513423.1 | 5 | 610 | |||
XM_011515122.2 | XP_011513424.1 | 4 | 566 | |||
XM_011515123.2 | XP_011513425.1 | 4 | 566 | |||
GenBank mRNA | BC031107.1 | - | 5 | 697 | ||
BC045737.1 | - | 4 | 550 | |||
BI560654.1 | - | 4 | 546 |